Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103656.3 | NP_001097126.1 | 5 - 6 | 2362 | 75 | |
NM_001298829.1 | NP_001285758.1 | 5 - 6 | 1553 | 75 | ||
NM_057539.5 | NP_476887.2 | 6 - 7 | 2042 | 75 | ||
NM_164833.2 | NP_723413.1 | 5 - 6 | 1501 | 75 | ||
NM_164834.2 | NP_723414.2 | 6 - 7 | 2611 | 75 | ||
GenBank mRNA | BT003533.1 | - | 6 - 7 | 2038 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |