Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: raw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201804.1 | NP_001188733.1 | 2 - 3 | 1013 | 69 | |
NM_001298831.1 | NP_001285760.1 | 1 - 2 | 1334 | 69 | ||
NM_164836.3 | NP_723416.1 | 2 - 3 | 813 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |