Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: raw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201804.1 | NP_001188733.1 | 6 - 7 | 1793 | 109 | |
NM_001298830.1 | NP_001285759.1 | 5 - 6 | 1243 | 109 | ||
NM_001298831.1 | NP_001285760.1 | 5 - 6 | 2114 | 109 | ||
NM_080050.3 | NP_524789.2 | 5 - 6 | 1059 | 109 | ||
NM_164836.3 | NP_723416.1 | 6 - 7 | 1593 | 109 | ||
GenBank mRNA | BT044207.1 | - | 5 - 6 | 1030 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |