Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: raw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201804.1 | NP_001188733.1 | 9 - 10 | 3246 | 70 | |
NM_001298830.1 | NP_001285759.1 | 8 - 9 | 2696 | 70 | ||
NM_001298831.1 | NP_001285760.1 | 8 - 9 | 3567 | 70 | ||
NM_080050.3 | NP_524789.2 | 8 - 9 | 2512 | 70 | ||
NM_164836.3 | NP_723416.1 | 9 - 10 | 3046 | 70 | ||
GenBank mRNA | BT044207.1 | - | 8 - 9 | 2483 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |