Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103663.1 | NP_001097133.1 | 2 - 3 | 740 | 90 | |
NM_001201828.2 | NP_001188757.1 | 8 - 9 | 3632 | 90 | ||
NM_001201829.2 | NP_001188758.1 | 8 - 9 | 3568 | 90 | ||
NM_001382120.1 | NP_001369124.1 | 9 - 10 | 4527 | 90 | ||
NM_078801.6 | NP_523525.2 | 8 - 9 | 3973 | 90 | ||
NM_164858.2 | NP_723468.1 | 8 - 9 | 3856 | 90 | ||
GenBank mRNA | AY119554.1 | - | 8 - 9 | 3856 | 90 | |
BT015212.1 | - | 2 - 3 | 740 | 90 | ||
KX531630.1 | - | 2 - 3 | 303 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |