Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259031.2 | NP_001245960.1 | 1 - 2 | 205 | 118 | |
NM_164885.3 | NP_723510.1 | 1 - 2 | 205 | 118 | ||
GenBank mRNA | BT089033.1 | - | 1 - 2 | 177 | 118 | |
BT099837.1 | - | 1 - 2 | 199 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |