Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bsk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169459.1 | NP_001162930.1 | 1 - 2 | 648 | 60 | |
NM_001169461.3 | NP_001162932.1 | 1 - 2 | 506 | 60 | ||
NM_164900.3 | NP_723541.1 | 2 - 3 | 386 | 60 | ||
GenBank mRNA | AY070865.1 | - | 2 - 3 | 263 | 60 | |
AY122221.1 | - | 2 - 3 | 365 | 60 | ||
U49180.1 | - | 1 - 2 | 649 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |