Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Utx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201844.1 | NP_001188773.1 | 11 - 12 | 2555 | 98 | |
NM_001273390.1 | NP_001260319.1 | 9 - 10 | 2286 | 98 | ||
NM_135524.4 | NP_609368.3 | 11 - 12 | 2573 | 98 | ||
NM_164907.4 | NP_723550.2 | 9 - 10 | 2108 | 98 | ||
NM_164908.4 | NP_723551.2 | 9 - 10 | 2096 | 98 | ||
GenBank mRNA | AY129437.1 | - | 11 - 12 | 2638 | 98 | |
BT010089.1 | - | 11 - 12 | 2573 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |