Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ir31a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273417.1 | NP_001260346.1 | 4 - 5 | 1917 | 80 | |
NM_164926.2 | NP_723585.2 | 4 - 5 | 1917 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |