Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fatp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169469.3 | NP_001162940.3 | 4 - 5 | 1186 | 57 | |
NM_001273425.3 | NP_001260354.1 | 2 - 3 | 813 | 57 | ||
NM_079984.4 | NP_524723.2 | 4 - 5 | 1186 | 57 | ||
NM_164934.2 | NP_723597.1 | 3 - 4 | 1262 | 57 | ||
GenBank mRNA | AY052105.1 | - | 4 - 5 | 1173 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |