Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ge-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135642.3 | NP_609486.2 | 4 - 5 | 1121 | 83 | |
NM_164957.2 | NP_723654.1 | 3 - 4 | 1204 | 83 | ||
GenBank mRNA | BT004479.1 | - | 4 - 5 | 1101 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |