Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ge-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135642.3 | NP_609486.2 | 6 - 7 | 2957 | 90 | |
NM_164957.2 | NP_723654.1 | 5 - 6 | 3040 | 90 | ||
GenBank mRNA | BT004479.1 | - | 6 - 7 | 2937 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |