Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crol | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259064.2 | NP_001245993.1 | 5 - 6 | 3363 | 64 | |
NM_001273461.1 | NP_001260390.1 | 6 - 7 | 2552 | 64 | ||
NM_001273462.1 | NP_001260391.1 | 4 - 5 | 2465 | 64 | ||
NM_001273464.1 | NP_001260393.1 | 6 - 7 | 2827 | 64 | ||
NM_057895.4 | NP_477243.1 | 6 - 7 | 3473 | 64 | ||
NM_057896.4 | NP_477244.1 | 6 - 7 | 2855 | 64 | ||
NM_164971.2 | NP_723687.1 | 6 - 7 | 2575 | 64 | ||
GenBank mRNA | AY119553.1 | - | 3 - 4 | 1893 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |