Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ir25a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273120.1 | NP_001260049.1 | 7 - 8 | 2410 | 74 | |
NM_001273121.1 | NP_001260050.1 | 6 - 7 | 1902 | 74 | ||
NM_001273122.1 | NP_001260051.1 | 7 - 8 | 2103 | 74 | ||
NM_135019.2 | NP_608863.2 | 8 - 9 | 1968 | 74 | ||
GenBank mRNA | HQ600588.1 | - | 7 - 8 | 1930 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |