Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hoe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273130.1 | NP_001260059.1 | 6 - 7 | 2104 | 72 | |
NM_001273131.1 | NP_001260060.1 | 6 - 7 | 2032 | 72 | ||
NM_001298678.1 | NP_001285607.1 | 5 - 6 | 1831 | 72 | ||
NM_135031.4 | NP_608875.1 | 5 - 6 | 1816 | 72 | ||
NM_135032.2 | NP_608876.1 | 5 - 6 | 1749 | 72 | ||
NM_164603.2 | NP_723024.2 | 6 - 7 | 1960 | 72 | ||
NM_175966.2 | NP_787980.1 | 6 - 7 | 2044 | 72 | ||
NM_175967.2 | NP_787981.1 | 6 - 7 | 2082 | 72 | ||
NM_175968.2 | NP_787982.1 | 6 - 7 | 2028 | 72 | ||
GenBank mRNA | AF145605.1 | - | 5 - 6 | 1795 | 72 | |
AY050571.1 | - | 6 - 7 | 2047 | 72 | ||
AY050572.1 | - | 5 - 6 | 1798 | 72 | ||
BT016131.1 | - | 6 - 7 | 2047 | 72 | ||
BT044574.1 | - | 5 - 6 | 1744 | 72 | ||
BT126019.1 | - | 3 - 4 | 1267 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |