Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hoe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298678.1 | NP_001285607.1 | 1 - 2 | 471 | 69 | |
NM_135032.2 | NP_608876.1 | 1 - 2 | 389 | 69 | ||
GenBank mRNA | BT044574.1 | - | 1 - 2 | 384 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |