Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp28d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135068.4 | NP_608912.1 | 5 - 6 | 1386 | 62 | |
GenBank mRNA | AY069176.1 | - | 5 - 6 | 1367 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |