Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oscillin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258965.2 | NP_001245894.1 | 6 - 7 | 1038 | 68 | |
NM_135094.2 | NP_608938.1 | 5 - 6 | 1048 | 68 | ||
GenBank mRNA | AY122220.1 | - | 5 - 6 | 1048 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |