Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: flr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140385.4 | NP_648642.1 | 1 - 2 | 136 | 98 | |
GenBank mRNA | AY061340.1 | - | 1 - 2 | 117 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |