Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spt20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038916.3 | NP_001034005.2 | 1 - 2 | 676 | 80 | |
NM_001300133.1 | NP_001287062.1 | 1 - 2 | 676 | 80 | ||
NM_140402.5 | NP_648659.2 | 1 - 2 | 676 | 80 | ||
GenBank mRNA | BT016098.1 | - | 645 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |