Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neurl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140431.3 | NP_648688.2 | 3 - 4 | 1690 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |