Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bbg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259826.1 | NP_001246755.1 | 3 - 4 | 2768 | 61 | |
NM_140448.3 | NP_648705.2 | 3 - 4 | 2752 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |