Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mop | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140465.3 | NP_648722.2 | 2 - 3 | 962 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |