Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sytbeta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140477.2 | NP_648734.1 | 7 - 8 | 2025 | 101 | |
NM_144170.2 | NP_652427.2 | 7 - 8 | 1927 | 101 | ||
GenBank mRNA | BT133101.1 | - | 7 - 8 | 1928 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |