Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eig71Ek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140540.3 | NP_648797.1 | 2 - 3 | 255 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |