Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275019.1 | NP_001261948.1 | 6 - 7 | 1373 | 69 | |
NM_001275020.1 | NP_001261949.1 | 6 - 7 | 1361 | 69 | ||
NM_001275021.1 | NP_001261950.1 | 6 - 7 | 1370 | 69 | ||
NM_001275022.1 | NP_001261951.1 | 6 - 7 | 1355 | 69 | ||
NM_140636.3 | NP_648893.1 | 6 - 7 | 1382 | 69 | ||
GenBank mRNA | AY058257.1 | - | 6 - 7 | 1349 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |