Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lasp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259869.2 | NP_001246798.1 | 3 - 4 | 961 | 77 | |
NM_001259870.2 | NP_001246799.1 | 3 - 4 | 961 | 77 | ||
NM_140655.3 | NP_648912.2 | 3 - 4 | 961 | 77 | ||
NM_168681.4 | NP_730192.2 | 3 - 4 | 961 | 77 | ||
GenBank mRNA | BT003801.1 | - | 3 - 4 | 516 | 77 |
Gene Symbol: CG43954 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275037.2 | NP_001261966.1 | 3 - 4 | 961 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |