Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGRP-SB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140660.4 | NP_648917.1 | 1 - 2 | 59 | 60 | |
GenBank mRNA | BT023240.1 | - | 1 - 2 | 58 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster Inventoried |