Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: beg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140693.3 | NP_648950.1 | 1 | 1087 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |