Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scaf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014589.4 | NP_001014589.1 | 2 - 3 | 312 | 126 | |
NM_001038939.2 | NP_001034028.2 | 2 - 3 | 312 | 126 | ||
NM_001169986.3 | NP_001163457.1 | 2 - 3 | 312 | 126 | ||
GenBank mRNA | BT006010.1 | - | 2 - 3 | 297 | 126 | |
BT031129.1 | - | 2 - 3 | 278 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |