Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UQCR-Q | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202195.2 | NP_001189124.1 | 2 - 3 | 269 | 73 | |
NM_001202196.2 | NP_001189125.1 | 3 - 4 | 469 | 73 | ||
NM_001275058.1 | NP_001261987.1 | 2 - 3 | 269 | 73 | ||
NM_140728.4 | NP_648985.1 | 3 - 4 | 498 | 73 | ||
GenBank mRNA | AY113252.1 | - | 3 - 4 | 498 | 73 | |
KX531481.1 | - | 1 - 2 | 172 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |