Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: qjt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275059.1 | NP_001261988.1 | 1 | 710 | 65 | |
NM_140730.4 | NP_648987.1 | 2 | 866 | 65 | ||
GenBank mRNA | BT124827.1 | - | 2 | 866 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |