Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jon74E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202197.1 | NP_001189126.2 | 1 - 2 | 258 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |