Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jon74E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202197.1 | NP_001189126.2 | 2 - 3 | 518 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |