Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYPT-75D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275088.1 | NP_001262017.1 | 4 - 5 | 1888 | 66 | |
NM_001275089.2 | NP_001262018.1 | 3 - 4 | 2361 | 66 | ||
NM_001275090.1 | NP_001262019.1 | 5 - 6 | 1714 | 66 | ||
NM_140792.2 | NP_649049.1 | 4 - 5 | 1673 | 66 | ||
GenBank mRNA | AY075426.1 | - | 4 - 5 | 1652 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |