Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Max | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259904.2 | NP_001246833.1 | 2 - 3 | 273 | 69 | |
NM_001300185.1 | NP_001287114.1 | 2 - 3 | 273 | 69 | ||
NM_140840.4 | NP_649097.1 | 2 - 3 | 273 | 69 | ||
GenBank mRNA | BT025211.1 | - | 2 - 3 | 281 | 69 | |
KX532135.1 | - | 2 - 3 | 114 | 69 | ||
U77369.1 | - | 2 - 3 | 163 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |