Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wnd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202203.1 | NP_001189132.1 | 4 - 5 | 2064 | 64 | |
NM_140880.3 | NP_649137.3 | 5 - 6 | 1791 | 64 | ||
NM_176362.2 | NP_788540.1 | 5 - 6 | 1714 | 64 | ||
NM_176363.3 | NP_788541.1 | 5 - 6 | 1919 | 64 | ||
GenBank mRNA | AY094787.1 | - | 5 - 6 | 1741 | 64 | |
BT100072.1 | - | 4 - 5 | 2063 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |