Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tey | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144509.3 | NP_001137981.1 | 1 - 2 | 385 | 77 | |
NM_140883.2 | NP_649140.1 | 1 - 2 | 385 | 77 | ||
GenBank mRNA | BT133456.1 | - | 1 - 2 | 384 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |