Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: schuy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140909.2 | NP_649166.1 | 1 - 2 | 625 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |