Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: obst-F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140929.1 | NP_649186.1 | 1 - 2 | 163 | 71 | |
GenBank mRNA | AY118614.1 | - | 1 - 2 | 163 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |