Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gogo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140943.3 | NP_649200.2 | 7 - 8 | 2868 | 89 | |
GenBank mRNA | BT009982.1 | - | 7 - 8 | 2866 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |