Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RhoBTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275178.1 | NP_001262107.1 | 9 - 10 | 1963 | 69 | |
NM_001275179.1 | NP_001262108.1 | 9 - 10 | 1701 | 69 | ||
NM_140959.2 | NP_649216.2 | 9 - 10 | 1963 | 69 | ||
GenBank mRNA | AF217287.1 | - | 9 - 10 | 1940 | 69 | |
AY051710.1 | - | 9 - 10 | 1930 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |