Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vha68-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259086.2 | NP_001246015.1 | 2 - 3 | 136 | 70 | |
NM_001273495.1 | NP_001260424.1 | 2 - 3 | 166 | 70 | ||
NM_001273496.1 | NP_001260425.1 | 2 - 3 | 220 | 70 | ||
NM_143747.3 | NP_652004.2 | 2 - 3 | 178 | 70 | ||
NM_165021.2 | NP_723775.1 | 2 - 3 | 156 | 70 | ||
NM_165022.2 | NP_723776.1 | 2 - 3 | 223 | 70 | ||
GenBank mRNA | BT124897.1 | - | 2 - 3 | 166 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |