Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kuz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014481.3 | NP_001014481.1 | 9 - 10 | 2459 | 61 | |
NM_001103689.2 | NP_001097159.2 | 9 - 10 | 2586 | 61 | ||
NM_001103690.3 | NP_001097160.2 | 9 - 10 | 2459 | 61 | ||
NM_057839.5 | NP_477187.1 | 9 - 10 | 2586 | 61 | ||
NM_165047.3 | NP_723825.1 | 9 - 10 | 2459 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |