Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CenG1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078838.3 | NP_523562.2 | 4 - 5 | 1264 | 56 | |
NM_165067.2 | NP_723849.1 | 4 - 5 | 1383 | 56 | ||
NM_165068.2 | NP_723850.1 | 3 - 4 | 455 | 56 | ||
GenBank mRNA | AY122158.1 | - | 4 - 5 | 1246 | 56 | |
BT053695.1 | - | 3 - 4 | 433 | 56 | ||
BT133428.1 | - | 4 - 5 | 1380 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |