Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mhc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259122.1 | NP_001246051.1 | 10 - 11 | 2446 | 96 | |
NM_165181.3 | NP_723999.1 | 10 - 11 | 2286 | 96 | ||
NM_165186.3 | NP_724004.1 | 10 - 11 | 2286 | 96 | ||
NM_165188.3 | NP_724006.1 | 10 - 11 | 2286 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |