Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: blot | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300164.1 | NP_001287093.1 | 5 - 6 | 794 | 65 | |
NM_079401.3 | NP_524125.2 | 5 - 6 | 1759 | 65 | ||
NM_170636.2 | NP_730263.1 | 5 - 6 | 1329 | 65 | ||
NM_170637.2 | NP_730264.1 | 4 - 5 | 632 | 65 | ||
NM_170638.2 | NP_730265.1 | 4 - 5 | 762 | 65 | ||
GenBank mRNA | BT033062.1 | - | 4 - 5 | 632 | 65 | |
BT133104.1 | - | 5 - 6 | 797 | 65 | ||
BT133440.1 | - | 5 - 6 | 1709 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |