Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: for | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001347740.1 | NP_001334731.1 | 1 - 2 | 986 | 99 | |
NM_058139.4 | NP_477487.1 | 3 - 4 | 2145 | 99 | ||
NM_205904.2 | NP_995626.1 | 3 - 4 | 2131 | 99 | ||
NM_205906.2 | NP_995628.1 | 2 - 3 | 1866 | 99 | ||
GenBank mRNA | AY061514.1 | - | 3 - 4 | 2130 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |