Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: for | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001347740.1 | NP_001334731.1 | 2 - 3 | 1152 | 65 | |
NM_001369953.1 | NP_001356892.1 | 3 - 4 | 732 | 65 | ||
NM_001369954.1 | NP_001356896.1 | 2 - 3 | 453 | 65 | ||
NM_058139.4 | NP_477487.1 | 4 - 5 | 2311 | 65 | ||
NM_205904.2 | NP_995626.1 | 4 - 5 | 2297 | 65 | ||
NM_205906.2 | NP_995628.1 | 3 - 4 | 2032 | 65 | ||
GenBank mRNA | AY061514.1 | - | 4 - 5 | 2296 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |