Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: for | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014464.2 | NP_001014464.1 | 4 - 5 | 2207 | 79 | |
NM_001169387.2 | NP_001162858.1 | 5 - 6 | 2087 | 79 | ||
NM_001347740.1 | NP_001334731.1 | 5 - 6 | 1757 | 79 | ||
NM_001369953.1 | NP_001356892.1 | 6 - 7 | 1337 | 79 | ||
NM_001369954.1 | NP_001356896.1 | 5 - 6 | 1058 | 79 | ||
NM_001369955.1 | NP_001356955.1 | 4 - 5 | 788 | 79 | ||
NM_058139.4 | NP_477487.1 | 7 - 8 | 2916 | 79 | ||
NM_058141.4 | NP_477489.1 | 5 - 6 | 2099 | 79 | ||
NM_058142.4 | NP_477490.1 | 4 - 5 | 2659 | 79 | ||
NM_134319.4 | NP_599146.1 | 5 - 6 | 2539 | 79 | ||
NM_205904.2 | NP_995626.1 | 7 - 8 | 2902 | 79 | ||
NM_205906.2 | NP_995628.1 | 6 - 7 | 2637 | 79 | ||
NM_205907.2 | NP_995629.1 | 5 - 6 | 2147 | 79 | ||
GenBank mRNA | AF132175.2 | - | 5 - 6 | 2070 | 79 | |
AY061514.1 | - | 7 - 8 | 2901 | 79 | ||
BT004881.1 | - | 5 - 6 | 2575 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |